Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p19ARF/CDKN2A Antibody (5-C3-1), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N20174PECY7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N20174PECY7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N20174PECY7 Supplier Novus Biologicals Supplier No. NB200174PECY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

p19ARF/CDKN2A Monoclonal specifically detects p19ARF/CDKN2A in Human, Mouse, Golden Syrian Hamster (Negative) samples. It is validated for Flow Cytometry, Knockout Validated.

Specifications

Antigen p19ARF
Applications Flow Cytometry, Immunoassay
Classification Monoclonal
Clone 5-C3-1
Conjugate PE-Cyanine7
Dilution Flow Cytometry, Knockout Validated
Formulation PBS with 0.05% Sodium Azide
Gene Alias ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, p16, p16-INK4, p16INK4a, p16-INK4a, P19, p19ARF, TP16
Gene Symbols CDKN2A
Host Species Rat
Immunogen A synthetic peptide containing amino acids 54-75 of the murine p19ARF. [UniProt No. Q64364]
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline ABC Transporters, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, Hypoxia, MAP Kinase Signaling, Neuroscience, p53 Pathway, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 12578.0
Target Species Human, Mouse, Golden Syrian Hamster
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.