Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p19ARF/CDKN2A Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB338901 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338901 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338901 Supplier Novus Biologicals Supplier No. NB200106PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

p19ARF/CDKN2A Polyclonal antibody specifically detects p19ARF/CDKN2A in Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)

Specifications

Antigen p19ARF/CDKN2A
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS
Gene Alias alternative reading frame, ARF, ARF-INK4a, CDK4I, CDKN2A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (p16, inhibits CDK4), cyclin-dependent kinase inhibitor 2A, isoform 3, cyclin-dependent kinase inhibitor 2A, isoforms 1/2, cyclin-dependent kinase inhibitor protein, INK4, INK4a, Ink4a/Arf, INK4a-ARF, mitochondrial smARF', MTS1, p14, p14ARF, P16, p16(INK4a), p16-INK4, p16INK4a, p16-INK4a, P19, p19<ARF>, p19ARF, Pctr1
Host Species Rabbit
Immunogen Synthetic peptide conjugated to KLH, derived from a sequence within residues 50-150 of mouse p19ARF/CDKN2A. [Swiss-Prot #Q64364]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Biologically Active Proteins, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, Hypoxia, MAP Kinase Signaling, Neuroscience, p53 Pathway, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 12578
Target Species Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.