Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p21/CIP1/CDKN1A Antibody (195720), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB297541 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297541 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297541 Supplier Novus Biologicals Supplier No. FAB1047V

Mouse Monoclonal Antibody

p21/CIP1/CDKN1A Monoclonal antibody specifically detects p21/CIP1/CDKN1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Flow Cytometry

Specifications

Antigen p21/CIP1/CDKN1A
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Flow Cytometry
Classification Monoclonal
Clone 195720
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CAP20cyclin-dependent kinase inhibitor 1, CDK-interacting protein 1, CDKN1melanoma differentiation associated protein 6, CIP1WAF1CDK-interaction protein 1, cyclin-dependent kinase inhibitor 1A (p21, Cip1), MDA6, MDA-6, Melanoma differentiation-associated protein 6, p21, p21CIP1, p21Cip1/Waf1, PIC1, SDI1DNA synthesis inhibitor, wild-type p53-activated fragment 1
Host Species Mouse
Immunogen E. coli-derived recombinant human p21/CIP1/CDKN1A, Ser2-Pro164, Accession # P38936
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1026
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.