Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p21/CIP1/CDKN1A Antibody (CIP1/4377R) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB119687 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119687 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119687 Supplier Novus Biologicals Supplier No. NBP308961
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

p21/CIP1/CDKN1A Monoclonal specifically detects p21/CIP1/CDKN1A in Human samples. It is validated for Immunohistochemistry-Paraffin.

Specifications

Antigen p21/CIP1/CDKN1A
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CIP1/4377R
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 1-2 ug/mL
Formulation 10mM PBS
Gene Alias CAP20cyclin-dependent kinase inhibitor 1, CDK-interacting protein 1, CDKN1melanoma differentiation associated protein 6, CIP1WAF1CDK-interaction protein 1, cyclin-dependent kinase inhibitor 1A (p21, Cip1), MDA6, MDA-6, Melanoma differentiation-associated protein 6, p21, p21CIP1, p21Cip1/Waf1, PIC1, SDI1DNA synthesis inhibitor, wild-type p53-activated fragment 1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to p21/CIP1/CDKN1A residues within aa1-100 of p21/CIP1/CDKN1A was used as an immunogen.
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1026
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.