Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p21/Waf1/Cip1 Rat anti-Human,Mouse, Unconjugated, Clone: HUGO-291, MilliporeSigma™
Rat Monoclonal Antibody
$182.31 - $461.76
Specifications
Antigen | p21/Waf1/Cip1 |
---|---|
Clone | HUGO-291 |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABE18162MI
|
MilliporeSigma
MABE181625UG |
25 μg |
Each for $182.31
|
|
|||||
MABE18161MI
|
MilliporeSigma
MABE1816100UG |
100 μg |
Each for $461.76
|
|
|||||
Description
p21/Waf1/Cip1 Monoclonal antibody specifically detects p21/Waf1/Cip1 in Human,Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blotting.Specifications
p21/Waf1/Cip1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rat | |
Human, Mouse | |
Cyclin-dependent kinase inhibitor 1;CDK-interacting protein 1;Melanoma differentiation-associated protein | |
GST/His-tagged full-length murine recombinant p21 protein. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼26 kDa observed; 17.78 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
HUGO-291 | |
Monoclonal | |
Purified | |
Epigenetics and Nuclear Function | |
NP_031695.1 | |
Cdkn1a, Cip1, Waf1 | |
IgG2a κ | |
Protein G Purified | |
Clone HUGO-291 is a rat monoclonal antibody that specifically detects p21 protein. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title