Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p23/PTGES3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP219999 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219999 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219999 Supplier Novus Biologicals Supplier No. NBP219999
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

p23/PTGES3 Polyclonal specifically detects p23/PTGES3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen p23/PTGES3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.31mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Accession No. Q15185
Gene Alias cPGESp23, Cytosolic prostaglandin E2 synthase, Hsp90 co-chaperone, P23cytosolic prostaglandin E synthase, Progesterone receptor complex p23, prostaglandin E synthase 3, prostaglandin E synthase 3 (cytosolic), TEBPEC 5.3.99.3, Telomerase-binding protein p23, unactive progesterone receptor, 23 kD
Gene Symbols PTGES3
Host Species Rabbit
Immunogen Full length human p23 Recombinant protein.
Molecular Weight of Antigen 19 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 10728
Test Specificity Human, Mouse, Rat. Species reactivity based on sequence identity: Bovine (100%), Rhesus Monkey (100%), Chimpanzee (100%).
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.