Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p23/PTGES3 Antibody (OTI2D2), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB273186A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB273186A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB273186A48 Supplier Novus Biologicals Supplier No. NBP273186AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p23/PTGES3 Monoclonal specifically detects p23/PTGES3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen p23/PTGES3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2D2
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cPGESp23, Cytosolic prostaglandin E2 synthase, Hsp90 co-chaperone, P23cytosolic prostaglandin E synthase, Progesterone receptor complex p23, prostaglandin E synthase 3, prostaglandin E synthase 3 (cytosolic), TEBPEC 5.3.99.3, Telomerase-binding protein p23, unactive progesterone receptor, 23 kD
Gene Symbols PTGES3
Host Species Mouse
Immunogen Full length recombinant protein of human p23/PTGES3 (NP_006592) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 10728.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.