Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p27/Kip1 Antibody (DCS-72.F6 + KIP1/769), PerCP, Novus Biologicals™
SDP

Catalog No. NB007336 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB007336 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB007336 Supplier Novus Biologicals Supplier No. NBP247771PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p27/Kip1 Monoclonal specifically detects p27/Kip1 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen p27/Kip1
Applications Flow Cytometry
Classification Monoclonal
Clone DCS-72.F6 + KIP1/769
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CDKN4, cyclin-dependent kinase inhibitor 1B, cyclin-dependent kinase inhibitor 1B (p27, Kip1), Cyclin-dependent kinase inhibitor p27, KIP1P27KIP1, MEN1B, MEN4, p27Kip1
Gene Symbols CDKN1B
Host Species Mouse
Immunogen Mouse recombinant p27/Kip1 protein (DCS-72.F6); Recombinant human CDKN1B protein (KIP1/769) (Uniprot: P46527)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Phospho Specific, Prostate Cancer, Tumor Suppressors
Primary or Secondary Primary
Test Specificity Recognizes a 27kDa protein, identified as the p27Kip1, a cell cycle regulatory mitotic inhibitor. Its epitope spans between aa 83-204 of p27. It is highly specific and shows no cross-reaction with other related mitotic inhibitors. p27Kip1 functions as a negative regulator of G1 progression and has been proposed to function as a possible mediator of TGF- induced G1 arrest. p27Kip1 is a candidate tumor suppressor gene. This monoclonal antibody co-precipitates cdk4 in complex p27Kip1 and is excellent for staining of formalin-fixed tissues.
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.