Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p27 Rat anti-Mouse, Unconjugated, Clone: SON82, MilliporeSigma™
Rat Monoclonal Antibody
$189.29 - $477.80
Specifications
Antigen | p27 |
---|---|
Clone | SON82 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABE18152MI
|
MilliporeSigma
MABE181525UG |
25 μg |
Each for $189.29
|
|
|||||
MABE18151MI
|
MilliporeSigma
MABE1815100UG |
100 μg |
Each for $477.80
|
|
|||||
Description
p27 Monoclonal antibody specifically detects p27 in Mouse samples. It is validated for Immunohistochemistry (Paraffin).Specifications
p27 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rat | |
Mouse | |
Cyclin-dependent kinase inhibitor 1B;Cyclin-dependent kinase inhibitor p27;p27Kip1 | |
His-tagged full-length recombinant murine p27 protein. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
22.19 kDa calculated. |
SON82 | |
Monoclonal | |
Purified | |
Epigenetics and Nuclear Function | |
NP_034005 | |
Cdkn1b | |
IgG2a κ | |
Protein G Purified | |
SON82 is a rat monoclonal antibody that specifically detects mouse p27Kip1 protein. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title