Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2X1/P2RX1 Antibody (551820) [DyLight 405], Novus Biologicals™
SDP

Catalog No. NB401422 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB401422 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB401422 Supplier Novus Biologicals Supplier No. FAB10533E
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

P2X1/P2RX1 Monoclonal antibody specifically detects P2X1/P2RX1 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen P2X1/P2RX1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 551820
Conjugate DyLight 405
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ATP receptor, P2X purinoceptor 1, P2X receptor, subunit 1, P2X1P2X1 receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 1
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived transfected with human P2X1/P2RX1, Accession # P51575
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5023
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.