Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2X3/P2RX3 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP2148380.025ML
Description
P2X3/P2RX3 Polyclonal specifically detects P2X3/P2RX3 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
P2X3/P2RX3 | |
Polyclonal | |
Unconjugated | |
Whole antisera with No Preservative | |
P2RX3 | |
A synthetic peptide from the cytoplasmic domain of mouse P2X3 conjugated to an immunogenic carrier protein was used as the antigen. | |
0.025 mL | |
Primary | |
Reconstitute with 0.1 ml sterilized water to desired concentration. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
LYOPH | |
Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:300-1:2000 | |
ATP receptor, MGC129956, P2X purinoceptor 3, P2X receptor, subunit 3, P2X3purinergic receptor P2X3, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 3, purinoceptor P2X3 | |
Goat | |
Unpurified | |
RUO | |
5024 | |
Rat, Mouse | |
Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction