Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2X3/P2RX3 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | P2X3/P2RX3 |
---|---|
Concentration | LYOPH |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
P2X3/P2RX3 Polyclonal specifically detects P2X3/P2RX3 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
P2X3/P2RX3 | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Goat | |
Rat, Mouse | |
5024 | |
IgG | |
Unpurified |
LYOPH | |
Polyclonal | |
Antisera | |
RUO | |
ATP receptor, MGC129956, P2X purinoceptor 3, P2X receptor, subunit 3, P2X3purinergic receptor P2X3, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 3, purinoceptor P2X3 | |
P2RX3 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title