Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2X5/P2RX5 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB052805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB052805 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB052805 Supplier Novus Biologicals Supplier No. NBP298199AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

P2X5/P2RX5 Polyclonal specifically detects P2X5/P2RX5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen P2X5/P2RX5
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ATP receptor, ATP receptor subunit, ionotropic ATP receptor P2X5, LRH-1, MGC47755, P2X purinoceptor 5, P2X5lymphoid-restricted histocompatibility antigen-1, P2X5R, Purinergic receptor, purinergic receptor P2X ligand gated ion channel 5, purinergic receptor P2X, ligand-gated ion channel, 5
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human P2X5/P2RX5 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5026
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.