Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2X6/P2RX6 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018484 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018484 0.1 mg
NBP130565 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018484 Supplier Novus Biologicals Supplier No. NBP1305650.1MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

P2X6/P2RX6 Polyclonal specifically detects P2X6/P2RX6 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.

Specifications

Antigen P2X6/P2RX6
Applications Immunohistochemistry (Free Floating), Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunohistochemistry Free-Floating
Formulation Sterilized water with No Preservative
Gene Alias ATP receptor, MGC129625, P2RXL1skeletal muscle-expressed P2X, P2X purinoceptor 6, P2X6purinoceptor P2X6, P2XMpurinoreceptor P2X6, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 6, Purinergic receptor P2X-like 1, purinergic receptor P2X-like 1, orphan receptor
Gene Symbols P2RX6
Host Species Rabbit
Immunogen A synthetic peptide from the extracellular domain of human P2X6 (P2RX6, P2X purinoceptor 6, ATP receptor, P2RXL1) conjugated to an immunogenic carrier protein was used as the immunogen. The antigen shares 90% identity with mouse sequence.
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9127
Test Specificity Appears to be specific for P2RX6.
Reconstitution Reconstitute with 0.5 ml sterilized water.
Target Species Rat, Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.