Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2X6/P2RX6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | P2X6/P2RX6 |
---|---|
Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
P2X6/P2RX6 Polyclonal antibody specifically detects P2X6/P2RX6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
P2X6/P2RX6 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Cardiovascular Biology, Endocrinology, Neuroscience, Signal Transduction | |
PBS (pH 7.2), 40% Glycerol | |
9127 | |
IgG | |
Immunogen affinity purified |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
ATP receptor, MGC129625, P2RXL1skeletal muscle-expressed P2X, P2X purinoceptor 6, P2X6purinoceptor P2X6, P2XMpurinoreceptor P2X6, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 6, Purinergic receptor P2X-like 1, purinergic receptor P2X-like 1, orphan receptor | |
This antibody was developed against a recombinant protein corresponding to amino acids: VTQIKELGNRLWDVADFVKPPQGENVFFLVTNFLVTPAQVQGRCPEHPSVPLAN | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title