Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2X7/P2RX7 Antibody (1058613), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB295370 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295370 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295370 Supplier Novus Biologicals Supplier No. FAB97903L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

P2X7/P2RX7 Monoclonal specifically detects P2X7/P2RX7 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen P2X7/P2RX7
Applications Flow Cytometry
Classification Monoclonal
Clone 1058613
Conjugate DyLight 550
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias ATP receptor, MGC20089, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A
Host Species Mouse
Immunogen Human P2X7/P2RX7 synthetic peptide, Accession # Q99572
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5027
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.