Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2X7/P2RX7 Antibody (CL11508), Novus Biologicals™
SDP

Catalog No. NB239929 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB239929 25 μg
NB239924 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB239929 Supplier Novus Biologicals Supplier No. NBP32119625UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

P2X7/P2RX7 Monoclonal antibody specifically detects P2X7/P2RX7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen P2X7/P2RX7
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL11508
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunohistochemistry 1:500- 1:1000, Immunohistochemistry-Paraffin 1:500- 1:1000
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias ATP receptor, MGC20089, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A
Host Species Mouse
Immunogen This antibody was developed against a Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence of Q99572. Actual immunogen sequence is proprietary.
Purification Method Protein A purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5027
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.