Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2X7/P2RX7 Antibody (CL11508), Novus Biologicals™

Mouse Monoclonal Antibody
$330.00 - $547.00
Specifications
Antigen | P2X7/P2RX7 |
---|---|
Clone | CL11508 |
Dilution | Western Blot 1 ug/ml, Immunohistochemistry 1:500- 1:1000, Immunohistochemistry-Paraffin 1:500- 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
P2X7/P2RX7 Monoclonal antibody specifically detects P2X7/P2RX7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
P2X7/P2RX7 | |
Western Blot 1 ug/ml, Immunohistochemistry 1:500- 1:1000, Immunohistochemistry-Paraffin 1:500- 1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ATP receptor, MGC20089, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A | |
This antibody was developed against a Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence of Q99572. Actual immunogen sequence is proprietary. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CL11508 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Immunology, Signal Transduction | |
PBS, pH 7.2, 40% glycerol | |
5027 | |
IgG2a | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title