Learn More
Description
Specifications
Specifications
| Antigen | P2X7/P2RX7 |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | Hano43 |
| Conjugate | DyLight 755 |
| Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence |
| Gene Alias | ATP receptor, MGC20089, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A |
| Gene Symbols | P2RX7 |
| Host Species | Rat |
| Immunogen | A P2X7-expression construct and a final boost with P2X7-transfected HEK cells. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
