Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2X7/P2RX7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP120180 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP120180 0.1 mL
NB018399 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP120180 Supplier Novus Biologicals Supplier No. NBP120180
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

P2X7/P2RX7 Polyclonal specifically detects P2X7/P2RX7 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen P2X7/P2RX7
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunohistochemistry 1:1000, Immunohistochemistry-Paraffin 1:1000
Formulation Lyophilized from whole antisera with No Preservative
Gene Accession No. Q9Z1M0
Gene Alias ATP receptor, MGC20089, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A
Gene Symbols P2RX7
Host Species Rabbit
Immunogen A synthetic peptide from mouse P2X7 conjugated to blue carrier protein was used as the antigen.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5027
Reconstitution Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size (0.025ml) is provided in reconstituted format.
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.