Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2X7/P2RX7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | P2X7/P2RX7 |
|---|---|
| Concentration | LYOPH |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
P2X7/P2RX7 Polyclonal specifically detects P2X7/P2RX7 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| P2X7/P2RX7 | |
| Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Rat, Monkey, Human, Mouse | |
| ATP receptor, MGC20089, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A | |
| P2RX7 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Q9Z1M0 | |
| 5027 | |
| IgG | |
| Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title