Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2Y10/P2RY10 Antibody (639329), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB315363 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB315363 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB315363 Supplier Novus Biologicals Supplier No. FAB10158PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

P2Y10/P2RY10 Monoclonal specifically detects P2Y10/P2RY10 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen P2Y10/P2RY10
Applications Flow Cytometry
Classification Monoclonal
Clone 639329
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias G-protein coupled purinergic receptor P2Y10, P2Y purinoceptor 10, P2Y10P2Y-like receptor, purinergic receptor P2Y, G-protein coupled, 10, putative P2Y purinoceptor 10
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human P2RY10, Accession # O00398
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers
Primary or Secondary Primary
Gene ID (Entrez) 27334
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.