Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2Y11/P2RY11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB166098 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166098 20 μg
NB166097 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166098 Supplier Novus Biologicals Supplier No. NBP31550120UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

P2Y11/P2RY11 Polyclonal antibody specifically detects P2Y11/P2RY11 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen P2Y11/P2RY11
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias P2Y11P2Y purinoceptor 11, purinergic receptor P2Y, G-protein coupled, 11, purinergic receptor P2Y11
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 300-379 of human P2Y11/P2RY11 (NP_002557.2).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers
Primary or Secondary Primary
Gene ID (Entrez) 5032
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.