Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2Y11/P2RY11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$159.60 - $389.00
Specifications
Antigen | P2Y11/P2RY11 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
P2Y11/P2RY11 Polyclonal antibody specifically detects P2Y11/P2RY11 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
P2Y11/P2RY11 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
P2Y11P2Y purinoceptor 11, purinergic receptor P2Y, G-protein coupled, 11, purinergic receptor P2Y11 | |
A synthetic peptide corresponding to a sequence within amino acids 300-379 of human P2Y11/P2RY11 (NP_002557.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers | |
PBS, 50% glycerol, pH7.3 | |
5032 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title