Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2Y12/P2RY12 Antibody, Novus Biologicals™
SDP

Catalog No. NB018756 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018756 0.025 mg
NBP17824905 0.05 mg
NBP178249 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB018756 Supplier Novus Biologicals Supplier No. NBP1782490.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

P2Y12/P2RY12 Polyclonal specifically detects P2Y12/P2RY12 in Human, Mouse, Rat, Porcine, Monkey, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen P2Y12/P2RY12
Applications Immunoprecipitation, Immunocytochemistry, Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:10000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:100, Dot Blot 1:10000
Formulation Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide
Gene Accession No. Q9H244
Gene Alias ADPG-R, Gi-coupled ADP receptor HORK3, HORK3P2Y12 platelet ADP receptor, P2T(AC), P2Y purinoceptor 12, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12ADP-glucose receptor, purinergic receptor P2RY12, purinergic receptor P2Y, G-protein coupled, 12, putative G-protein coupled receptor, SP1999G-protein coupled receptor SP1999
Gene Symbols P2RY12
Host Species Rabbit
Immunogen Synthetic peptide selected from human P2Y12 receptor gene corresponding to amino acids with the 320-342 at the C-terminus of protein. The peptide was covalently modified to achieve desired antigenicity before coupling to a carrier protein.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 64805
Test Specificity This antibody detects a single 56-68kDa P2Y12 diffused band. The antibody does not cross reacts with other P2X or with other P2Y receptors on western blots.
Target Species Monkey, Primate, Mouse, Human, Pig, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.