Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2Y12/P2RY12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
$702.91
Specifications
| Antigen | P2Y12/P2RY12 |
|---|---|
| Dilution | Western Blot - reported in scientific literature (PMID:31968618)., Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence -validated from a verified customer review, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Immunohistochemistry-Frozen -validated from a verified customer review |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
P2Y12/P2RY12 Polyclonal specifically detects P2Y12/P2RY12 in Human, Canine, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| P2Y12/P2RY12 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| Human, Feline | |
| Q9H244 | |
| 64805 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KSFRNSLISMLKCPNSATSLSQDNRKKEQDGGDPNEETPM | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot - reported in scientific literature (PMID:31968618)., Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence -validated from a verified customer review, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Immunohistochemistry-Frozen -validated from a verified customer review | |
| Polyclonal | |
| Rabbit | |
| GPCR | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ADPG-R, Gi-coupled ADP receptor HORK3, HORK3P2Y12 platelet ADP receptor, P2T(AC), P2Y purinoceptor 12, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12ADP-glucose receptor, purinergic receptor P2RY12, purinergic receptor P2Y, G-protein coupled, 12, putative G-protein coupled receptor, SP1999G-protein coupled receptor SP1999 | |
| P2RY12 | |
| IgG | |
| Affinity Purified | |
| Specificity of human P2Y12/P2RY12 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title