Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2Y12/P2RY12 Antibody (SR1887), Novus Biologicals™
SDP

Catalog No. NB258190 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258190 100 μg
NB259193 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB258190 Supplier Novus Biologicals Supplier No. NBP322528100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

P2Y12/P2RY12 Monoclonal antibody specifically detects P2Y12/P2RY12 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen P2Y12/P2RY12
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone SR1887
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias ADPG-R, Gi-coupled ADP receptor HORK3, HORK3P2Y12 platelet ADP receptor, P2T(AC), P2Y purinoceptor 12, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12ADP-glucose receptor, purinergic receptor P2RY12, purinergic receptor P2Y, G-protein coupled, 12, putative G-protein coupled receptor, SP1999G-protein coupled receptor SP1999
Host Species Rabbit
Immunogen A synthesized peptide derived from human P2Y12/P2RY12 (Uniprot #: Q9H244)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 64805
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.