Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2Y12/P2RY12 Antibody (SR1887), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | P2Y12/P2RY12 |
---|---|
Clone | SR1887 |
Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
P2Y12/P2RY12 Monoclonal antibody specifically detects P2Y12/P2RY12 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
P2Y12/P2RY12 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
ADPG-R, Gi-coupled ADP receptor HORK3, HORK3P2Y12 platelet ADP receptor, P2T(AC), P2Y purinoceptor 12, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12ADP-glucose receptor, purinergic receptor P2RY12, purinergic receptor P2Y, G-protein coupled, 12, putative G-protein coupled receptor, SP1999G-protein coupled receptor SP1999 | |
A synthesized peptide derived from human P2Y12/P2RY12 (Uniprot #: Q9H244) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1887 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
GPCR | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
64805 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title