Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2Y12 Receptor, Rabbit anti-Rat, Human, Polyclonal, Millipore Sigma™
Rabbit Polyclonal Antibody
$192.34 - $482.80
Specifications
Antigen | P2Y12 Receptor |
---|---|
Dilution | Immunohistochemistry (Paraffin) Analysis: A 1:250-1,000 dilution from a representative lot detected P2Y12 Receptor in human cerebral cortex and human tonsil tissue sections.Western Blotting Analysis: 1μg/mL from a representative lot detected P2Y12 Receptor in human platelet cell lysate. |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABS148725UG
|
MilliporeSigma
ABS148725UG |
25 μg |
Each for $192.34
|
|
|||||
ABS1487100
|
MilliporeSigma
ABS1487100UG |
100 μg |
Each for $482.80
|
|
|||||
Description
Anti-P2Y12 Receptor, Cat. No. ABS1487, is a rabbit polyclonal antibody that detects P2Y purinoceptor 12 and has been tested for Immunohistochemistry (Paraffin) and Western Blotting.
P2Y purinoceptor 12 (UniProt: Q9H244; also known as P2Y12, ADP-glucose receptor, ADPG-R, P2T(AC), P2Y(AC), P2Y(cyc), P2Y12 platelet ADP receptor, P2Y(ADP), SP1999) is encoded by the P2RY12 (also known as HORK3) gene (Gene ID: 64805) in human. P2Y12 is a multi-pass metabotropic receptor that is highly expressed in the platelets. Lower expression levels have been observed in the brain, lung, and adrenal glands. P2Y12 serves as a receptor for ADP and ATP coupled to G-proteins that inhibit the adenylyl cyclase second messenger system. It is not activated by UDP and UTP. It is required for normal platelet aggregation and blood coagulation. It is the predominant receptor involved in the ADP-stimulated activation of the glycoprotein IIb/IIIa receptor. Activation of the glycoprotein IIb/IIIa receptor results in enhanced platelet degranulation and thromboxane production, and prolonged platelet aggregation. P2Y12 receptor has also been reported to be expressed on microglia and is required to neuropathic pain following peripheral nerve injury. P2Y12 has seven transmembrane, four cytoplasmic, and four extracellular domains. The transmembrane helices are somewhat tilted and/or kinked. Agonist binding promotes a conformation change in the extracellular loops that leads to an inward movement of the transmembrane helices. Mutations in P2RY12 gene have been linked to bleeding disorders resulting from severe impairment of platelet response to ADP resulting in defective platelet aggregation. (Ref.: Burnstock G. (2009). Curr. Pharm. Des. 15(15);1717-35; Damman, P., et al. (2012). J. Thromb. Thrombolysis. 33(2); 143-153).Specifications
P2Y12 Receptor | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
RUO | |
Rat, Human | |
P2Y purinoceptor 12;ADP-glucose receptor;ADPG-R;P2T(AC);P2Y(AC);P2Y(cyc);P2Y12 platelet ADP receptor;P2Y(ADP);SP1999 | |
P2RY12;HORK3 | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
∽50 kDa observed; 39.44 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Immunohistochemistry (Paraffin) Analysis: A 1:250-1,000 dilution from a representative lot detected P2Y12 Receptor in human cerebral cortex and human tonsil tissue sections.Western Blotting Analysis: 1μg/mL from a representative lot detected P2Y12 Receptor in human platelet cell lysate. | |
Polyclonal | |
Rabbit | |
Signaling | |
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NP_073625 | |
KLH-conjugated linear peptide corresponding to 13 amino acids from the internal region of human P2Y purinoreceptor 12. | |
Affinity Purified | |
This rabbit polyclonal antibody specifically detects P2Y12 receptor. It targets an epitope with in 13 amino acids from the extracellular domain. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title