Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p34-Arc/ARPC2 Rabbit anti-Human,Bovine,Mouse,Rat, KHL, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$189.29 - $481.52
Specifications
Antigen | p34-Arc/ARPC2 |
---|---|
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | KLH |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
07227I25UMI
|
MilliporeSigma
07227I25UG |
25 μg |
Each for $189.29
|
|
|||||
07227I100MI
|
MilliporeSigma
07227I100UG |
100 μg |
Each for $481.52
|
|
|||||
Description
p34-Arc/ARPC2 Polyclonal antibody specifically detects p34-Arc/ARPC2 in Human,Bovine,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.Specifications
p34-Arc/ARPC2 | |
Polyclonal | |
Purified | |
Cell Structure | |
NP_005722 | |
ARPC2, ARC34, PRO2446 | |
IgG | |
Affinity Purified | |
Anti-Anti-p34-Arc\ARPC2 is a rabbit polyclonal antibody that detects Actin-related protein 2/3 complex subunit 2. It targets an epitope with in 14 amino acids from the C-terminal region. |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
KLH | |
Rabbit | |
Human, Bovine, Mouse, Rat | |
Actin-related protein 2/3 complex subunit 2;Arp2/3 complex 34 kDa subunit;p34-ARC | |
KLH-conjugated linear peptide corresponding to 14 amino acids from the C-terminal region of human Actin-related protein 2/3 complex subunit 2 (p34-ARC). | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼34 kDa observed; 34.33 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title