Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p35 baculovirus Antibody, Novus Biologicals™
SDP

Catalog No. NB10056153T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056153T 0.025 mL
NB10056153 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056153T Supplier Novus Biologicals Supplier No. NB10056153SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 9 publications

p35 baculovirus Polyclonal specifically detects p35 baculovirus in Bombyx mori (Silkworm), Drosophila, Silkworm, Virus samples. It is validated for IF/IHC, Immunocytochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen p35 baculovirus
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 21514385), Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen Assay Dependent, Immunocytochemistry
Formulation Neat whole antisera with 0.05% Sodium Azide
Gene Alias p35 Baculovirus, p35-Baculovirus
Host Species Rabbit
Immunogen Full-length recombinant p35 baculovirus protein was used as immunogen.
Molecular Weight of Antigen 34 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Test Specificity p35 baculovirus may be observed as one or multiple bands on western blot: p35 baculovirus is cleaved during its reaction with caspases. This results in cleavage of p35; 10 and 25 kDa fragments have been described. Additional breakdown p35 products may also result.
Target Species Silkworm, Drosophila, Silkworm, Virus
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.