Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p38 alpha Antibody (0004), Novus Biologicals™
SDP

Catalog No. NB287377 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287377 20 μg
NB287554 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB287377 Supplier Novus Biologicals Supplier No. NBP32350720UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

p38 alpha Monoclonal antibody specifically detects p38 alpha in Human samples. It is validated for Western Blot

Specifications

Antigen p38 alpha
Applications Western Blot
Classification Monoclonal
Clone 4
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias CSBP;CSBP1;CSBP2;CSPB1;EXIP;MAPK14;Mitogen-activated protein kinase;Mitogen-activated protein kinase 14;Mxi2;p38;p38ALPHA;PRKM14;PRKM15;RK;SAPK2A
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with a synthetic phosphopeptide corresponding to residues around Thr180, Tyr182 of human phospho-p38 alpha.
Purification Method Protein A purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Immunology, Innate Immunity, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1432
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.