Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p38 alpha Antibody (0004), Novus Biologicals™

Rabbit Monoclonal Antibody
$393.00
Specifications
Antigen | p38 alpha |
---|---|
Clone | 4 |
Dilution | Western Blot 1:1000-1:10000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
p38 alpha Monoclonal antibody specifically detects p38 alpha in Human samples. It is validated for Western BlotSpecifications
p38 alpha | |
Western Blot 1:1000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CSBP;CSBP1;CSBP2;CSPB1;EXIP;MAPK14;Mitogen-activated protein kinase;Mitogen-activated protein kinase 14;Mxi2;p38;p38ALPHA;PRKM14;PRKM15;RK;SAPK2A | |
This antibody was obtained from a rabbit immunized with a synthetic phosphopeptide corresponding to residues around Thr180, Tyr182 of human phospho-p38 alpha. | |
Protein A purified |
4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Cell Cycle and Replication, Immunology, Innate Immunity, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction | |
0.2 um filtered solution in PBS | |
1432 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title