Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p38 alpha Antibody (9F12), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB11096875R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11096875R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11096875R Supplier Novus Biologicals Supplier No. NB11096875R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p38 alpha Monoclonal specifically detects p38 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry.

Specifications

Antigen p38
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone 9F12
Conjugate DyLight 550
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. NP_001306.1
Gene Alias CSAID-binding protein, Csaids binding protein, CSBP, CSBP1MAX-interacting protein 2, CSBP2, CSPB1, cytokine suppressive inflammatory drug binding protein, Cytokine suppressive inflammatory drug-binding protein, EC 2.7.11, EC 2.7.11.24, EXIP, MAP kinase 14, MAP kinase MXI2, MAP kinase p38 alpha, MAPK 14, mitogen-activated protein kinase 14, Mitogen-activated protein kinase p38 alpha, Mxi2, p38, p38 MAP kinase, p38 mitogen activated protein kinase, p38ALPHA, p38alpha Exip, PRKM14, PRKM15, RK, SAPK2A, stress-activated protein kinase 2A
Gene Symbols MAPK14
Host Species Mouse
Immunogen Full length recombinant protein expressed in E.coli cells
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Cycle and Replication, Immunology, Innate Immunity, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1432
Test Specificity Detects approx 38kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.