Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p38 beta/MAPK11 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB105928 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB105928 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB105928 Supplier Novus Biologicals Supplier No. NBP298810JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

p38 beta/MAPK11 Polyclonal specifically detects p38 beta/MAPK11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen p38 beta/MAPK11
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50 mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.24, MAP kinase 11, MAP kinase p38 beta, MAPK 11, mitogen-activated protein kinase 11, Mitogen-activated protein kinase p38 beta, mitogen-activated protein kinase p38-2, p38b, p38Beta, P38BETA2, PRKM11, SAPK2B, SAPK2p38-2P38B, Stress-activated protein kinase 2, stress-activated protein kinase-2, stress-activated protein kinase-2b
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human p38 beta/MAPK11.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5600
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.