Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p38 beta/MAPK11 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056446T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056446T 0.025 mg
NB10056446 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056446T Supplier Novus Biologicals Supplier No. NB10056446SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

p38 beta/MAPK11 Polyclonal specifically detects p38 beta/MAPK11 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.

Specifications

Antigen p38 beta/MAPK11
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. NP_002742
Gene Alias EC 2.7.11, EC 2.7.11.24, MAP kinase 11, MAP kinase p38 beta, MAPK 11, mitogen-activated protein kinase 11, Mitogen-activated protein kinase p38 beta, mitogen-activated protein kinase p38-2, p38b, p38Beta, P38BETA2, PRKM11, SAPK2B, SAPK2p38-2P38B, Stress-activated protein kinase 2, stress-activated protein kinase-2, stress-activated protein kinase-2b
Gene Symbols MAPK11
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a portion amino acids 1-100 was used as immunogen,
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5600
Test Specificity In human or mouse brain, a 38 kDa band is observed.
Target Species Human, Mouse, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.