Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p38 delta/SAPK4 Antibody (OTI12B2), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB302129 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302129 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302129 Supplier Novus Biologicals Supplier No. NBP273188APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p38 delta/SAPK4 Monoclonal antibody specifically detects p38 delta/SAPK4 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen p38 delta/SAPK4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone OTI12B2
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias EC 2.7.11, EC 2.7.11.24, MAP kinase 13, MAPK 13, MGC99536, mitogen-activated protein kinase 13, Mitogen-activated protein kinase p38 delta, p38delta, PRKM13, SAPK4MAP kinase p38 delta, Stress-activated protein kinase 4
Host Species Mouse
Immunogen Full length human recombinant protein of human MAPK13(NP_002745) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5603
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.