Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p38 delta/SAPK4 Antibody (OTI12B2), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP273188G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273188G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273188G Supplier Novus Biologicals Supplier No. NBP273188G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p38 delta/SAPK4 Monoclonal specifically detects p38 delta/SAPK4 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen p38 delta/SAPK4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI12B2
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.11, EC 2.7.11.24, MAP kinase 13, MAPK 13, MGC99536, mitogen-activated protein kinase 13, Mitogen-activated protein kinase p38 delta, p38delta, PRKM13, SAPK4MAP kinase p38 delta, Stress-activated protein kinase 4
Gene Symbols MAPK13
Host Species Mouse
Immunogen Full length recombinant protein of human p38 delta/SAPK4 (NP_002745) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5603.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.