Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p38 gamma/SAPK3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15827020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15827020 20 μL
NBP158270 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15827020 Supplier Novus Biologicals Supplier No. NBP15827020UL

Rabbit Polyclonal Antibody

p38 gamma/SAPK3 Polyclonal specifically detects p38 gamma/SAPK3 in Human samples. It is validated for Western Blot.

Specifications

Antigen p38 gamma/SAPK3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P53778
Gene Alias EC 2.7.11, ERK3, ERK-6, ERK6MAPK 12, Extracellular signal-regulated kinase 6, MAP kinase 12, MAP kinase p38 gamma, mitogen-activated protein kinase 12, mitogen-activated protein kinase 3, Mitogen-activated protein kinase p38 gamma, P38GAMMA, PRKM12, SAPK-3, SAPK3EC 2.7.11.24, Stress-activated protein kinase 3
Gene Symbols MAPK12
Host Species Rabbit
Immunogen Synthetic peptides corresponding to MAPK12(mitogen-activated protein kinase 12) The peptide sequence was selected from the N terminal of MAPK12. Peptide sequence SGAYGAVCSAVDGRTGAKVAIKKLYRPFQSELFAKRAYRELRLLKHMRHE.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6300
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.