Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p38 gamma/SAPK3 Antibody (OTI10E1), PerCP, Novus Biologicals™
SDP

Catalog No. NB273189PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB273189PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB273189PCP Supplier Novus Biologicals Supplier No. NBP273189PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p38 gamma/SAPK3 Monoclonal specifically detects p38 gamma/SAPK3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen p38 gamma/SAPK3
Applications Flow Cytometry
Classification Monoclonal
Clone OTI10E1
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 2.7.11, ERK3, ERK-6, ERK6MAPK 12, Extracellular signal-regulated kinase 6, MAP kinase 12, MAP kinase p38 gamma, mitogen-activated protein kinase 12, mitogen-activated protein kinase 3, Mitogen-activated protein kinase p38 gamma, P38GAMMA, PRKM12, SAPK-3, SAPK3EC 2.7.11.24, Stress-activated protein kinase 3
Gene Symbols MAPK12
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human p38 gamma/SAPK3 expression vector.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6300.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.