Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ P3NP Monoclonal Antibody (C2)
Mouse Monoclonal Antibody
$243.00 - $588.50
Specifications
Antigen | P3NP |
---|---|
Clone | C2 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541663
|
Invitrogen™
MA541663 |
200 μL |
Each for $588.50
|
|
|||||
PIMA543877
|
Invitrogen™
MA543877 |
20 μL |
Each for $243.00
|
|
|||||
Description
P3NP Monoclonal Antibody for Western Blot
Procollagen type III N-terminal peptide (P3NP) is released as part of collagen synthesis process in muscle. The increase of circulating P3NP is a marker not only of muscle growth, but also of muscle repair and fibrosis. P3NP may be a potential biomarker for sarcopenia.Specifications
P3NP | |
1 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P02461 | |
1281 | |
Recombinant protein Procollagen III N-Terminal Propeptide. | |
Antibody |
C2 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2 κ | |
Human | |
COL3A1 | |
PIIINP gene; Procollagen 3 N-Terminal Propeptide; Procollagen III N-Terminal Propeptide | |
COL3A1 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title