Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p40/deltaNp63 Antibody (P40/4396R), PerCP, Novus Biologicals™
SDP

Catalog No. NB187176 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB187176 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB187176 Supplier Novus Biologicals Supplier No. NBP314050PCP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

p40/deltaNp63 Monoclonal antibody specifically detects p40/deltaNp63 in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen p40/deltaNp63
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone P40/4396R
Conjugate PerCP
Formulation PBS with 0.05% Sodium Azide
Gene Alias AIS, amplified in squamous cell carcinoma, B(p51A), B(p51B), Chronic ulcerative stomatitis protein, EEC3, Keratinocyte transcription factor KET, KET, LMS, OFC8, p53CP, SHFM4, TP53CP, TP53L, TP63, TP73L, Transformation-related protein 63, tumor protein p53-competing protein
Host Species Rabbit
Immunogen Recombinant fragment (around aa1-100) of human p40/deltaNp63 protein (exact sequence is proprietary) (Uniprot: Q9H3D4 )
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8626
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.