Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p400 Antibody, Novus Biologicals™
SDP

Catalog No. NBP219663 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219663 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219663 Supplier Novus Biologicals Supplier No. NBP219663
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

p400 Polyclonal specifically detects p400 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen p400
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.95mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q96L91
Gene Alias CAG repeat protein 32, CAGH32hDomino, DKFZP434I225, Domino homolog, E1A binding protein p400, E1A-binding protein p400, EC 3.6.1, EC 3.6.4.-, FLJ45115, KIAA1498p400 kDa SWI2/SNF2-related protein, KIAA1818, P400, p400 SWI2/SNF2-related protein, TNRC12FLJ42018, trinucleotide repeat containing 12, Trinucleotide repeat-containing gene 12 protein
Gene Symbols EP400
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human p400. The exact sequence is proprietary.
Molecular Weight of Antigen 343 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 57634
Test Specificity Human. Species reactivity based on sequence identity: Mouse (84%), Rat (83%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.