Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p41 Flagellin Antibody (6802), FITC, Novus Biologicals™
SDP

Catalog No. NB390075 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390075 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390075 Supplier Novus Biologicals Supplier No. NBP308849F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p41 Flagellin Monoclonal antibody specifically detects p41 Flagellin in Bacteria samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen p41 Flagellin
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 6802
Conjugate FITC
Formulation PBS
Gene Alias 41 kDa antigen, fla, Flagellar filament 41 kDa core protein, Flagellin, p41
Host Species Mouse
Immunogen A crude sonicate of Borrelia spirochetes
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 46847386
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.