Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega P450-Glo™ CYP3A4 Assay with Luciferin-IPA
Click to view available options
Quantity:
10 mL
50 mL
Description
Includes
Luciferin Detection Reagent; P450-Glo Buffer; 5mM Luciferin-CEE, -ME, -H, -BE, -PFBE, -PPXE, or IPA
- Luminescent Assays and Screening Systems for Measuring CYP3A4 Activity
- Screening systems include complete set of reagents
- Luminescent format eliminates need for time-consuming analyses
- Broad dynamic range and low background
Specifications
Specifications
Quantity | 50 mL |
Content And Storage | -30°C to -10°C |
Detection Method | Homogeneous, Luminescence |
For Use With (Application) | For measuring cytochrome P450 activity |
Kit Contents | 1 x 1 Each Luciferin detection reagent, 1 x 50mL Reconstitution Buffer with Esterase, 1 x 60μL Luciferin-IPA |
Product Type | P450-Glo CYP3A4 Assay with Luciferin-IPA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction