Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p47 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989437
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9437 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9437 Supplier Bioss Supplier No. BS9902R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Major protein kinase C substrate of platelets.

Specifications

Antigen p47
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PLEK
Gene Accession No. P08567
Gene Alias 2010300B13Rik; FLJ27168; P47; Platelet 47 kDa protein; Pleckstrin; Plek
Gene Symbols PLEK
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Pleckstrin/p47.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5341
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.