Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 AIP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP176742 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176742 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176742 Supplier Novus Biologicals Supplier No. NBP176742
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

p53 AIP1 Polyclonal specifically detects p53 AIP1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry.

Specifications

Antigen p53 AIP1
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 4-8 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 ug/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. BAB16421
Gene Alias p53AIP1, p53-regulated apoptosis-inducing protein 1, tumor protein p53 regulated apoptosis inducing protein 1
Gene Symbols TP53AIP1
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human p53AIP1. The immunogen is located within the first 50 amino acids of p53AIP1. Amino Acid Squence: GSSSEASFRSAQASC
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 63970
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.