| Antigen |
p53 |
| Applications |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification |
Monoclonal |
| Clone |
BP53-12 + DO-7 |
| Conjugate |
FITC |
| Dilution |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Formulation |
PBS with 0.05% Sodium Azide |
| Gene Alias |
Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53 |
| Gene Symbols |
TP53 |
| Host Species |
Mouse |
| Immunogen |
Recombinant human wild-type p53 protein (BP53-12) & Recombinant human wild type p53 protein expressed in E. coli (DO-7) (Uniprot: P04637) |
| Purification Method |
Protein A or G purified |
| Quantity |
0.1 mL |
| Regulatory Status |
RUO |
| Research Discipline |
Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors |
| Primary or Secondary |
Primary |
| Test Specificity |
Recognizes a 53kDa protein, which is identified as p53 suppressor gene product. It reacts with the mutant as well as the wild form of p53 under denaturing and non-denaturing conditions. Its epitope maps within the N-terminus (aa 20-25) of p53 oncoprotein. p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to MDM2, SV40 T antigen and human papilloma virus E6 protein. Positive nuclear staining with p53 antibody has been reported to be a negative prognostic factor in breast carcinoma, lung carcinoma, colorectal, and urothelial carcinoma. Anti-p53 positivity has also been used to differentiate uterine serous carcinoma from endometrioid carcinoma as well as to detect intratubular germ cell neoplasia. Mutations involving p53 are found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma. |
| Target Species |
Human, Bovine, Primate |
| Content And Storage |
Store at 4°C in the dark. |
| Isotype |
IgG2a κ, IgG2b κ |
|
Show More
Show Less
|
|