Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB200171646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200171646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB200171646 Supplier Novus Biologicals Supplier No. NB200171JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

p53 Polyclonal specifically detects p53 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence, Knockdown Validated.

Specifications

Antigen p53
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, KnockDown
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence, Knockdown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53
Gene Symbols TP53
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residues 50-100 of human tumor protein p53 using the SwissProt entry P04637 (GeneID 7157).
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7157
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.