Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p53 Antibody (PAb 240) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 31 publication
$206.50 - $499.50
Specifications
Antigen | p53 |
---|---|
Clone | PAb 240 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:1000-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:250-1:500, Immunocytochemistry/Immunofluorescence 5 ug/ml. Use reported in scientific literature (PMID 1394225), Immunoprecipitation 10ug/mg, Immunohistochemistry-Paraffin 1:250-1:500, Immunohistochemistry-Frozen reported in multiple pieces of scientific literature, Flow (Intracellular), CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB200103SS
![]() |
Novus Biologicals
NB200103SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB200103
![]() |
Novus Biologicals
NB200103 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
p53 Monoclonal specifically detects p53 in Human, Mouse, Rat, Yeast, Xenopus (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready.Specifications
p53 | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Hypoxia, p53 Pathway, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53 | |
TP53 | |
IgG1 κ | |
Protein G purified | |
This monoclonal recognizes both mutant forms and wild-type human p53 under denaturing conditions. It does not recognize nondenatured wild type p53. In nondenaturing conditions (IP) it only recognizes mutant p53. |
PAb 240 | |
Western Blot 1:1000-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:250-1:500, Immunocytochemistry/Immunofluorescence 5 ug/ml. Use reported in scientific literature (PMID 1394225), Immunoprecipitation 10ug/mg, Immunohistochemistry-Paraffin 1:250-1:500, Immunohistochemistry-Frozen reported in multiple pieces of scientific literature, Flow (Intracellular), CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Yeast, Xenopus (Negative) | |
7157 | |
Gel-purified p53-beta-galactosidase fusion protein containing murine p53 from aa 14-389 (derived from pSV53C cDNA clone). | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title